2-Nitro-4-(trifluoromethyl)benzhydrazide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC300715-1G
APOSPC300715-1GEA 140 GBP
APOSPC300715-1G
2-Nitro-4-(trifluoromethyl)benzhydrazide
2-Nitro-4-(trifluoromethyl)benzhydrazide
Formula: C₈H₆F₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18804172
CAS Number: 830334-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR