3-(Cycloheptylamino)-1,1,1-trifluoropropan-2-ol, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC28101-1G
APOSPC28101-1GEA 1140 GBP
APOSPC28101-1G
3-(Cycloheptylamino)-1,1,1-trifluoropropan-2-ol, tech
3-(Cycloheptylamino)-1,1,1-trifluoropropan-2-ol
Formula: C₁₀H₁₈F₃NO
MDL Number: MFCD02083028
CAS Number: 478050-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR