6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC28097-1G
APOSPC28097-1GEA 1140 GBP
APOSPC28097-1G
6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile, tech
6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile
Formula: C₁₂H₁₂F₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD01316507
CAS Number: 478049-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR