(3E)-1,1,1-Trifluoro-4-[(2-fluorophenyl)amino]but-3-en-2-one, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC28052-1G
APOSPC28052-1GEA 1140 GBP
APOSPC28052-1G
(3E)-1,1,1-Trifluoro-4-[(2-fluorophenyl)amino]but-3-en-2-one, tech
(3E)-1,1,1-Trifluoro-4-[(2-fluorophenyl)amino]but-3-en-2-one
Formula: C₁₀H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD02186438
CAS Number: 380430-76-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR