4-(2-Fluorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC28040-1G
APOSPC28040-1GEA 1140 GBP
APOSPC28040-1G
4-(2-Fluorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech
4-(2-Fluorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide
Formula: C₁₄H₁₃FN₂O₂
MW: 260.264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00141882
CAS Number: 338404-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR