4-Bromo-2-fluorobenzyl fluoride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC27931-1G PC27931-250MG
APOSPC27931-1GEA 441 GBP
APOSPC27931-1G APOSPC27931-250MG
4-Bromo-2-fluorobenzyl fluoride 95%
4-Bromo-2-fluorobenzyl fluoride
Formula: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD31664908
CAS Number: 2244086-30-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR