2,3-Difluorobenzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC2654-10G PC2654-250G PC2654-50G PC2654-25G PC2654-100G PC2654-500G
APOSPC2654-10GEA 13.7 GBP
APOSPC2654-10G APOSPC2654-250G APOSPC2654-50G APOSPC2654-25G APOSPC2654-100G APOSPC2654-500G
2,3-Difluorobenzoic acid 98%
2,3-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00010267
CAS Number: 4519-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR