1,1-Dibromo-3,3,3-trifluoroacetone 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

PC2326-100G PC2326-500G
APOSPC2326-100GEA 75.6 GBP
APOSPC2326-100G APOSPC2326-500G
1,1-Dibromo-3,3,3-trifluoroacetone 95%
1,1-Dibromo-3,3,3-trifluoroacetone

Treatment with sodium acetate readily forms trifluoropyruvaldehyde hydrate, a versatile intermediatesee: J. Org. Chem., 58, 4033 (1993).

Formula: Br₂HCC(O)CF₃
Boiling Pt: 111…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041362
CAS Number: 431-67-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR