2,5-Dibromo-α,α,α-trifluorotoluene 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

PC2195-100G PC2195-25G PC2195-5G
APOSPC2195-100GEA 56.7 GBP
APOSPC2195-100G APOSPC2195-25G APOSPC2195-5G
2,5-Dibromo-α,α,α-trifluorotoluene 98%
2,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
Melting Pt: 46…48 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013554
CAS Number: 7657-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR