4-Bromo-2,5-difluorobenzaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC210045-1G PC210045-5G PC210045-25G
APOSPC210045-1GEA 15.8 GBP
APOSPC210045-1G APOSPC210045-5G APOSPC210045-25G
4-Bromo-2,5-difluorobenzaldehyde
4-Bromo-2,5-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD11847047
CAS Number: 357405-75-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR