N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC210004-1G
APOSPC210004-1GEA 306 GBP
APOSPC210004-1G
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide
Formula: C₁₅H₁₁ClF₃N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD28023539
CAS Number: 1610047-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR