2-Chloro-5-(trifluoromethyl)phenyl isocyanate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-Chloro-2-isocyanato-4-(trifluoromethyl)benzene

PC2074NE-1G
APOSPC2074NE-1GEA 16.8 GBP
APOSPC2074NE-1G
2-Chloro-5-(trifluoromethyl)phenyl isocyanate 98%
2-Chloro-5-(trifluoromethyl)phenyl isocyanate

  • May form small amount of sediment.

Formula: C₈H₃ClF₃NO
Boiling Pt: 63…64 °C/5mmHg
Density: 1.48 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037029
CAS Number: 50528-86-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR