5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC201071-1G
APOSPC201071-1GEA 69.3 GBP
APOSPC201071-1G
5-Chloro-3-(trifluoromethyl)pyridin-2-ol 97%
5-Chloro-3-(trifluoromethyl)pyridin-2-ol
Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20275098
CAS Number: 1214342-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR