(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride 96%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC201004-1G PC201004-250MG PC201004-5G
APOSPC201004-1GEA 194 GBP
APOSPC201004-1G APOSPC201004-250MG APOSPC201004-5G
(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride 96%
(4-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride
Formula: C₇H₇F₃N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD04117763
CAS Number: 1005515-26-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR