2-Chloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetohydrazide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200322-1G PC200322-500MG
APOSPC200322-1GEA 136 GBP
APOSPC200322-1G APOSPC200322-500MG
2-Chloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetohydrazide
2-Chloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetohydrazide
Formula: C₈H₆Cl₂F₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD18377864
CAS Number: 1383468-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR