3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200208-1G PC200208-500MG
APOSPC200208-1GEA 389 GBP
APOSPC200208-1G APOSPC200208-500MG
3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 95%
3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₉H₄F₃N₃O₄
Melting Pt: 172 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD12827808
CAS Number: 1186405-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR