Potassium-4-(4-morpholinylcarbonyl)phenyltrifluoroborate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200137-1G PC200137-500MG PC200137-5G
APOSPC200137-1GEA 38 GBP
APOSPC200137-1G APOSPC200137-500MG APOSPC200137-5G
Potassium-4-(4-morpholinylcarbonyl)phenyltrifluoroborate
Potassium 4-(4-morpholinylcarbonyl)phenyltrifluoroborate
Formula: C₁₁H₁₂BF₃KNO₂
Storage Temperature: Ambient
MDL Number: MFCD09800742
CAS Number: 1314957-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR