4,6-Dioxo-9-(trifluoromethyl)-2,4,5,6-tetrahydro-1H-benzo[g]imidazo[1,2,3-ij]pteridin-12-ium chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200098-500MG
APOSPC200098-500MGEA 390 GBP
APOSPC200098-500MG
4,6-Dioxo-9-(trifluoromethyl)-2,4,5,6-tetrahydro-1H-benzo[g]imidazo[1,2,3-ij]pteridin-12-ium chloride
4,6-Dioxo-9-(trifluoromethyl)-2,4,5,6-tetrahydro-1H-benzo[g]imidazo[1,2,3-ij]pteridin-12-ium chloride
Formula: C₁₃H₈F₃N₄O₂·Cl
Storage Temperature: Ambient
MDL Number: MFCD25948722
CAS Number: 353245-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR