Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200071-1G PC200071-500MG PC200071-5G
APOSPC200071-1GEA 195 GBP
APOSPC200071-1G APOSPC200071-500MG APOSPC200071-5G
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Formula: C₉H₈F₃NO₄
MW: 251.16 g/mol
Melting Pt: 217…219 °C
Storage Temperature: Ambient
MDL Number: MFCD09881353
CAS Number: 103900-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR