2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC200050-1G PC200050-250MG
APOSPC200050-1GEA 18.9 GBP
APOSPC200050-1G APOSPC200050-250MG
2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine
2-Chloro-5-fluoro-4-(methylsulphanyl)pyrimidine
Formula: C₅H₄ClFN₂S
Storage Temperature: Ambient
MDL Number: MFCD16876993
CAS Number: 87789-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR