2-Chloro-4-fluorobenzylamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC1864B-25G PC1864B-100G
APOSPC1864B-25GEA 27.3 GBP
APOSPC1864B-25G APOSPC1864B-100G
2-Chloro-4-fluorobenzylamine 97%
2-Chloro-4-fluorobenzylamine
Formula: C₇H₇ClFN
Boiling Pt: 94…96 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042532
CAS Number: 15205-11-5
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR