2-Chloro-4-fluorobenzonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC1861L-100G PC1861L-25G PC1861L-250G
APOSPC1861L-100GEA 104 GBP
APOSPC1861L-100G APOSPC1861L-25G APOSPC1861L-250G
2-Chloro-4-fluorobenzonitrile 95%
2-Chloro-4-fluorobenzonitrile
Formula: C₇H₃ClFN
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR