2-Chloro-6-fluorobenzal chloride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC1826-250MG
APOSPC1826-250MGEA 110 GBP
APOSPC1826-250MG
2-Chloro-6-fluorobenzal chloride 98%
2-Chloro-6-fluorobenzal chloride
Formula: C₇H₄Cl₃F
Boiling Pt: 228…230 °C
Density: 1.446 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 62476-62-4
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR