2-Chloro-4'-fluoroacetanilide 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC1688F-100G PC1688F-25G PC1688F-5G
APOSPC1688F-100GEA 139 GBP
APOSPC1688F-100G APOSPC1688F-25G APOSPC1688F-5G
2-Chloro-4'-fluoroacetanilide 97%
2-Chloro-4'-fluoroacetanilide
Formula: C₈H₇ClFNO
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00015730
CAS Number: 351-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR