2-Chloro-4-fluoroanisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC1623-100G PC1623-25G PC1623-5G
APOSPC1623-100GEA 214 GBP
APOSPC1623-100G APOSPC1623-25G APOSPC1623-5G
2-Chloro-4-fluoroanisole 98%
2-Chloro-4-fluoroanisole
Formula: C₇H₆ClFO
Boiling Pt: 197…200 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00672965
CAS Number: 2267-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR