4-Bromo-2-(trifluoromethoxy)benzenesulphonylchloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Bromo-2-(trifluoromethoxy)benzene-1-sulphonyl chloride

PC1593R-100G PC1593R-25G PC1593R-5G
APOSPC1593R-100GEA 172 GBP
APOSPC1593R-100G APOSPC1593R-25G APOSPC1593R-5G
4-Bromo-2-(trifluoromethoxy)benzenesulphonylchloride 97%
4-Bromo-2-(trifluoromethoxy)benzenesulphonylchloride
Formula: C₇H₃BrClF₃O₃S
Boiling Pt: 255…256 °C
Density: 1.85 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00179336
CAS Number: 175278-14-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR