5-Fluoro-2-methoxybenzonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC14564-1G PC14564-25G PC14564-5G
APOSPC14564-1GEA 15.8 GBP
APOSPC14564-1G APOSPC14564-25G APOSPC14564-5G
5-Fluoro-2-methoxybenzonitrile
5-Fluoro-2-methoxybenzonitrile
Formula: C₈H₆FNO
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD02683503
CAS Number: 189628-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR