(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

PC1208-100G PC1208-25G PC1208-500G
APOSPC1208-100GEA 26.3 GBP
APOSPC1208-100G APOSPC1208-25G APOSPC1208-500G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Peptide coupling reagent.

Formula: C₁₈H₂₈F₆N₆OP₂
MW: 520.4 g/mol
Melting Pt: 154…156 °C (dec.)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR