2-Morpholin-4-yl-2-[4-(trifluoromethyl)phenyl]ethylamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC1158-1G PC1158-250MG
APOSPC1158-1GEA 384 GBP
APOSPC1158-1G APOSPC1158-250MG
2-Morpholin-4-yl-2-[4-(trifluoromethyl)phenyl]ethylamine
2-Morpholin-4-yl-2-[4-(trifluoromethyl)phenyl]ethylamine
Formula: C₁₃H₁₇F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD01631947
CAS Number: 885950-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR