3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC110001-1G PC110001-250MG
APOSPC110001-1GEA 361 GBP
APOSPC110001-1G APOSPC110001-250MG
3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid
3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₆FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD05664782
CAS Number: 874800-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR