4-Nitro-2-(trifluoromethyl)aniline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine

PC1080-100G PC1080-10G PC1080-25G
APOSPC1080-100GEA 31.5 GBP
APOSPC1080-100G APOSPC1080-10G APOSPC1080-25G
4-Nitro-2-(trifluoromethyl)aniline 95%
4-Nitro-2-(trifluoromethyl)aniline
Formula: C₇H₅F₃N₂O₂
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR