N-(4-Fluorophenyl)rhodanine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC10583-1G
APOSPC10583-1GEA 67.3 GBP
APOSPC10583-1G
N-(4-Fluorophenyl)rhodanine
N-(4-Fluorophenyl)rhodanine
Formula: C₉H₆FNOS₂
Storage Temperature: Ambient
CAS Number: 387-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR