4-Bromo-2-(trifluoromethyl)aniline 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene

PC1042-100G PC1042-500G PC1042-50G
APOSPC1042-100GEA 36.8 GBP
APOSPC1042-100G APOSPC1042-500G APOSPC1042-50G
4-Bromo-2-(trifluoromethyl)aniline 98%
4-Bromo-2-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
Boiling Pt: 84…86 °C/5mmHg
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR