Pentafluorophenyl-2,5-dichlorobenzenesulphonate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC10323-1G PC10323-500MG PC10323-5G
APOSPC10323-1GEA 199 GBP
APOSPC10323-1G APOSPC10323-500MG APOSPC10323-5G
Pentafluorophenyl-2,5-dichlorobenzenesulphonate
Pentafluorophenyl 2,5-dichlorobenzenesulphonate
Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD05975133
CAS Number: 885950-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR