2-[(2-Fluorophenyl)amino]benzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-(2-Fluoroanilino)benzoic acid, N-(2-Fluorophenyl)anthranilic acid

PC0937-1G PC0937-5G
APOSPC0937-1GEA 60.9 GBP
APOSPC0937-1G APOSPC0937-5G
2-[(2-Fluorophenyl)amino]benzoic acid 98%
2-[(2-Fluorophenyl)amino]benzoic acid
Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00662257
CAS Number: 54-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR