Tris(perfluorobutyl)amine Mass spectrometry

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

PC0568-10X5G PC0568-5G PC0568-5X5G
APOSPC0568-10X5GEA 82 GBP
APOSPC0568-10X5G APOSPC0568-5G APOSPC0568-5X5G
Tris(perfluorobutyl)amine Mass spectrometry
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 178…180 °C
Melting Pt: -55
Density: 1.88 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR