3,4,5-Trifluoro-DL-phenylalanine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC0531-1G PC0531-5G PC0531-10G
APOSPC0531-1GEA 42 GBP
APOSPC0531-1G APOSPC0531-5G APOSPC0531-10G
3,4,5-Trifluoro-DL-phenylalanine
3,4,5-Trifluoro-DL-phenylalanine
Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01631414
CAS Number: 261952-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR