2-Methyl-5-nitrobenzotrifluoride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC0422-25G PC0422-5G PC0422-100G
APOSPC0422-25GEA 42 GBP
APOSPC0422-25G APOSPC0422-5G APOSPC0422-100G
2-Methyl-5-nitrobenzotrifluoride
2-Methyl-5-nitrobenzotrifluoride
Formula: C₈H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01631684
CAS Number: 89976-12-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR