3-(Trifluoromethyl)-p-anisidine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(Trifluoromethyl)-4-anisidine, 5-Amino-2-methoxybenzotrifluoride, 4-Methoxy-3-(trifluoromethyl)aniline, 3-(Trifluoromethyl)-4-methoxyaniline, 4-Amino-2-(trifluoromethyl)anisole

PC0397-1G PC0397-5G PC0397-10G
APOSPC0397-1GEA 15.8 GBP
APOSPC0397-1G APOSPC0397-5G APOSPC0397-10G
3-(Trifluoromethyl)-p-anisidine 98%
3-(Trifluoromethyl)-p-anisidine
Formula: C₈H₈F₃NO
Boiling Pt: 112 °C/18mmHg
Melting Pt: 58…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00043460
CAS Number: 393-15-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR