4,5-Difluoro-o-phenylenediamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4,5-Difluoro-1,2-phenylenediamine, 4,5-Difluorobenzene-1,2-diamine, 1,2-Diamino-4,5-difluorobenzene

PC0342-25G PC0342-100G PC0342-5G
APOSPC0342-25GEA 78.7 GBP
APOSPC0342-25G APOSPC0342-100G APOSPC0342-5G
4,5-Difluoro-o-phenylenediamine
4,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
Melting Pt: 131…132 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061131
CAS Number: 76179-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR