2,2-Bis(trifluoromethyl)propanoyl fluoride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Methyl-3,3,3-trifluoro-2-(trifluoromethyl)propanoyl fluoride, 2,2-Bis(trifluoromethyl)propionyl fluoride

PC0203-1G PC0203-25G PC0203-5G
APOSPC0203-1GEA 117 GBP
APOSPC0203-1G APOSPC0203-25G APOSPC0203-5G
2,2-Bis(trifluoromethyl)propanoyl fluoride
2,2-Bis(trifluoromethyl)propanoyl fluoride
Formula: C₅H₃F₇O
Boiling Pt: 47…49 °C
Density: 1.473 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236084
CAS Number: 1735-87-1
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR