4-Vinylbenzyl hexafluoroisopropyl ether

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC0184-1G PC0184-5G
APOSPC0184-1GEA 58.9 GBP
APOSPC0184-1G APOSPC0184-5G
4-Vinylbenzyl hexafluoroisopropyl ether
4-Vinylbenzyl hexafluoroisopropyl ether
Formula: C₁₂H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD00080684
CAS Number: 111158-92-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR