3-Chloro-4-(trifluoromethoxy)benzyl bromide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-(Bromomethyl)-2-chloro-1-(trifluoromethoxy)benzene

PC0181-1G PC0181-5G PC0181-25G PC0181-100G
APOSPC0181-1GEA 27.3 GBP
APOSPC0181-1G APOSPC0181-5G APOSPC0181-25G APOSPC0181-100G
3-Chloro-4-(trifluoromethoxy)benzyl bromide 98%
3-Chloro-4-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrClF₃O
MW: 289.476 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631560
CAS Number: 261763-18-2
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR