1-Bromo-2-(difluoromethyl)benzene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Bromobenzal fluoride

PC01660-100G PC01660-25G PC01660-5G
APOSPC01660-100GEA 435 GBP
APOSPC01660-100G APOSPC01660-25G APOSPC01660-5G
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
Formula: C₇H₅BrF₂
MW: 207.0154 g/mol
Boiling Pt: 62…63 °C/9mmHg
Storage Temperature: Ambient
MDL Number: MFCD06657962
CAS Number: 845866-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR