2-Chloro-5-nitrophenol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR9957-100G
APOSOR9957-100GEA 85.6 GBP
APOSOR9957-100G
2-Chloro-5-nitrophenol 97%
2-Chloro-5-nitrophenol
Formula: C₆H₄ClNO₃
Melting Pt: 112…119 °C
Storage Temperature: Ambient
MDL Number: MFCD01571825
CAS Number: 619-10-3
EINECS: 210-579-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR