Ethyl-4-morpholinobenzenecarboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR9853-1G OR9853-500MG OR9853-5G
APOSOR9853-1GEA 121 GBP
APOSOR9853-1G APOSOR9853-500MG APOSOR9853-5G
Ethyl-4-morpholinobenzenecarboxylate
Ethyl-4-morpholinobenzenecarboxylate
Formula: C₁₃H₁₇NO₃
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD06204958
CAS Number: 19614-15-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR