2-Bromo-(5-bromo-1-benzofuran-2-yl)ethan-1-one 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR9719-1G OR9719-250MG
APOSOR9719-1GEA 197 GBP
APOSOR9719-1G APOSOR9719-250MG
2-Bromo-(5-bromo-1-benzofuran-2-yl)ethan-1-one 97%
2-Bromo-(5-bromo-1-benzofuran-2-yl)ethan-1-one
Formula: C₁₀H₆Br₂O₂
Melting Pt: 135…136 °C
Storage Temperature: Ambient
MDL Number: MFCD06658982
CAS Number: 7039-76-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR