3-Nitrobenzophenone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (3-Nitrophenyl)(phenyl)methanone, m-Nitrobenzophenone

OR9685-25G OR9685-100G
APOSOR9685-25GEA 153 GBP
APOSOR9685-25G APOSOR9685-100G
3-Nitrobenzophenone 98%
3-Nitrobenzophenone
Formula: C₁₃H₉NO₃
MW: 227.22 g/mol
Melting Pt: 92…94 °C
Density: 1.367
Storage Temperature: Ambient
MDL Number: MFCD00007257
CAS Number: 2243-80-3
EINECS: 218-819-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR