4-(4-Chloropyrimidin-2-yl)morpholine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR965548-1G
APOSOR965548-1GEA 241 GBP
APOSOR965548-1G
4-(4-Chloropyrimidin-2-yl)morpholine 95%
4-(4-Chloropyrimidin-2-yl)morpholine
Formula: C₈H₁₀ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD00834977
CAS Number: 24192-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR