z-Tyr-OMe

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR963410-100G OR963410-25G
APOSOR963410-100GEA 68.3 GBP
APOSOR963410-100G APOSOR963410-25G
z-Tyr-OMe
z-Tyr-OMe
Formula: C₁₈H₁₉NO₅
Melting Pt: 93…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038784
CAS Number: 13512-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR